Product Details
- SNP ID
-
rs3765319
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:763650 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCCCTGTGTCCCCAGGAGGCTGC[A/G]CCCCTGGACGGAGTCCTGGCCAACC
- Phenotype
-
MIM: 601051
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR662
PubMed Links
Gene Details
- Gene
- MIR662
- Gene Name
- microRNA 662
There are no transcripts associated with this gene.
- Gene
- MSLN
- Gene Name
- mesothelin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001177355.2 |
316 |
Silent Mutation |
GCA,GCG |
A46A |
NP_001170826.1 |
NM_005823.5 |
316 |
Silent Mutation |
GCA,GCG |
A46A |
NP_005814.2 |
NM_013404.4 |
316 |
Silent Mutation |
GCA,GCG |
A46A |
NP_037536.2 |
XM_005255034.4 |
316 |
Silent Mutation |
GCA,GCG |
A45A |
XP_005255091.1 |
XM_011522348.2 |
316 |
Silent Mutation |
GCA,GCG |
A46A |
XP_011520650.1 |
XM_017022853.1 |
316 |
Silent Mutation |
GCA,GCG |
A56A |
XP_016878342.1 |
XM_017022854.1 |
316 |
Silent Mutation |
GCA,GCG |
A55A |
XP_016878343.1 |
XM_017022855.1 |
316 |
Silent Mutation |
GCA,GCG |
A56A |
XP_016878344.1 |
XM_017022856.1 |
316 |
Silent Mutation |
GCA,GCG |
A55A |
XP_016878345.1 |
XM_017022857.1 |
316 |
Silent Mutation |
GCA,GCG |
A46A |
XP_016878346.1 |
XM_017022858.1 |
316 |
Silent Mutation |
GCA,GCG |
A45A |
XP_016878347.1 |
View Full Product Details