Product Details

SNP ID
rs3786871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38265274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACCAAGAAGGCGTGGCACTGCC[C/T]CTTCCTGTGGCAAGAACGGGGTGAG
Phenotype
MIM: 608153 MIM: 605124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R14A PubMed Links

Gene Details

Gene
PPP1R14A
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14A
There are no transcripts associated with this gene.

Gene
SPINT2
Gene Name
serine peptidase inhibitor, Kunitz type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166103.1 Intron NP_001159575.1
NM_021102.3 Intron NP_066925.1

View Full Product Details