Product Details

SNP ID
rs3773749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138656522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAACTAAATAGAAATTCTATGA[C/T]GAAAGGCTATTACATTTTTAATAAT
Phenotype
MIM: 602925
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIK3CB PubMed Links

Gene Details

Gene
PIK3CB
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256045.1 Intron NP_001242974.1
NM_006219.2 Intron NP_006210.1
XM_005247530.2 Intron XP_005247587.1
XM_006713659.3 Intron XP_006713722.1
XM_011512895.2 Intron XP_011511197.1
XM_011512896.1 Intron XP_011511198.1
XM_017006619.1 Intron XP_016862108.1
XM_017006620.1 Intron XP_016862109.1
XM_017006621.1 Intron XP_016862110.1

View Full Product Details