Product Details

SNP ID
rs3783732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64389076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCTGAGAATATTCTGCATTCC[A/T]CTTGACCTCTGCCTGTTCTACCCTT
Phenotype
MIM: 172460
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MTHFD1 PubMed Links

Gene Details

Gene
MTHFD1
Gene Name
methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005956.3 Intron NP_005947.3

View Full Product Details