Product Details

SNP ID
rs3792914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176387701 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGGGGCCAGGCGGATCACTTGA[A/G]GCCCAGCCTGGCCAACATAGCGAAA
Phenotype
MIM: 118970 MIM: 612861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL10 PubMed Links

Gene Details

Gene
ARL10
Gene Name
ADP ribosylation factor like GTPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317948.1 Intron NP_001304877.1
NM_173664.5 Intron NP_775935.1
XM_011534529.2 Intron XP_011532831.1
XM_011534530.2 Intron XP_011532832.1
XM_011534531.2 Intron XP_011532833.1
XM_017009372.1 Intron XP_016864861.1
Gene
CLTB
Gene Name
clathrin light chain B
There are no transcripts associated with this gene.

Gene
HIGD2A
Gene Name
HIG1 hypoxia inducible domain family member 2A
There are no transcripts associated with this gene.

Gene
NOP16
Gene Name
NOP16 nucleolar protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256539.3 Intron NP_001243468.2
NM_001256540.3 Intron NP_001243469.2
NM_001291305.2 Intron NP_001278234.1
NM_001291307.2 Intron NP_001278236.1
NM_001291308.2 Intron NP_001278237.1
NM_001317975.1 Intron NP_001304904.1
NM_016391.7 Intron NP_057475.2
XM_011534567.1 Intron XP_011532869.1

View Full Product Details