Product Details

SNP ID
rs3814045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46512018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTTAGTGGTTCAACACTAGCTTC[A/G]CTTCCCAGAGGCTTATATAAAGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1E2 PubMed Links

Gene Details

Gene
ATP6V1E2
Gene Name
ATPase H+ transporting V1 subunit E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318063.1 2139 UTR 3 NP_001304992.1
XM_005264633.2 2139 UTR 3 XP_005264690.1
XM_005264634.2 2139 UTR 3 XP_005264691.1
XM_011533148.2 2139 UTR 3 XP_011531450.1
XM_011533149.2 2139 UTR 3 XP_011531451.1
XM_011533150.2 2139 UTR 3 XP_011531452.1
XM_011533151.2 2139 UTR 3 XP_011531453.1
XM_011533152.2 2139 UTR 3 XP_011531454.1
XM_011533153.2 2139 UTR 3 XP_011531455.1
XM_017005225.1 2139 UTR 3 XP_016860714.1
XM_017005226.1 2139 UTR 3 XP_016860715.1

View Full Product Details