Product Details

SNP ID
rs3794637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89687162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTTGTTTGTGGGGAGAATTTACA[C/T]CCGCGACGAGTTCGAGCTTGAAGGC
Phenotype
MIM: 603464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK10 PubMed Links

Gene Details

Gene
CDK10
Gene Name
cyclin dependent kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098533.2 358 Intron NP_001092003.2
NM_001160367.1 358 Intron NP_001153839.1
NM_052987.3 358 Intron NP_443713.2
NM_052988.4 358 Intron NP_443714.3
XM_006721308.2 358 Intron XP_006721371.1
XM_006721310.2 358 Intron XP_006721373.1
XM_011523405.2 358 Intron XP_011521707.1
XM_011523406.2 358 Intron XP_011521708.1
XM_011523407.2 358 Intron XP_011521709.1
XM_011523408.2 358 Intron XP_011521710.1
XM_011523410.2 358 Intron XP_011521712.1
XM_011523411.2 358 Intron XP_011521713.1
XM_011523412.1 358 Intron XP_011521714.1
XM_011523413.2 358 UTR 5 XP_011521715.1
XM_011523414.2 358 UTR 5 XP_011521716.1
XM_011523415.2 358 Intron XP_011521717.1
XM_011523416.2 358 Intron XP_011521718.1
XM_011523417.2 358 Intron XP_011521719.1
XM_017023806.1 358 Intron XP_016879295.1
XM_017023807.1 358 Intron XP_016879296.1
XM_017023808.1 358 Intron XP_016879297.1
XM_017023809.1 358 Intron XP_016879298.1
XM_017023810.1 358 UTR 5 XP_016879299.1
Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
There are no transcripts associated with this gene.

View Full Product Details