Product Details

SNP ID
rs3815136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47785126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTGGCCCTGGTCTAGGGCTGT[A/C]GCTAGAGATGGAGGCCAGTCCCTAC
Phenotype
MIM: 606542 MIM: 612187
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HDAC7 PubMed Links

Gene Details

Gene
HDAC7
Gene Name
histone deacetylase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098416.3 Intron NP_001091886.1
NM_001308090.1 Intron NP_001295019.1
NM_015401.4 Intron NP_056216.2
XM_011538478.1 Intron XP_011536780.1
XM_011538479.1 Intron XP_011536781.1
XM_011538480.1 Intron XP_011536782.1
XM_011538481.1 Intron XP_011536783.1
XM_011538482.1 Intron XP_011536784.1
XM_011538483.1 Intron XP_011536785.1
XM_017019455.1 Intron XP_016874944.1
XM_017019456.1 Intron XP_016874945.1
Gene
SLC48A1
Gene Name
solute carrier family 48 member 1
There are no transcripts associated with this gene.

View Full Product Details