Product Details

SNP ID
rs3734672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13787346 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAGTCAAGCCTTTCAACCGAGGT[A/G]CTTGGCATACTTAACGGAAAAAGCG
Phenotype
MIM: 616952
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCUR1 PubMed Links

Gene Details

Gene
MCUR1
Gene Name
mitochondrial calcium uniporter regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031713.3 2517 UTR 3 NP_001026883.1
XM_011514802.1 2517 UTR 3 XP_011513104.1

View Full Product Details