Product Details

SNP ID
rs3735240
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100122735 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTGAAATGGCAGAAAGGGGAAGG[A/G]AGGGGTGGGGTGGTGAGCTGGGCTG
Phenotype
MIM: 610047 MIM: 602955
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CNPY4 PubMed Links
Additional Information
For this assay, SNP(s) [rs76702812] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CNPY4
Gene Name
canopy FGF signaling regulator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152755.1 Intron NP_689968.1
Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
There are no transcripts associated with this gene.

Gene
MBLAC1
Gene Name
metallo-beta-lactamase domain containing 1
There are no transcripts associated with this gene.

Gene
TAF6
Gene Name
TATA-box binding protein associated factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190415.1 Intron NP_001177344.1
NM_005641.3 Intron NP_005632.1
NM_139315.2 Intron NP_647476.1

View Full Product Details