Product Details
- SNP ID
-
hCV27489472
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:75915316 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCCTCGGGCCTGGGCACTGCGC[A/G]TCTGGCCCGACGCTCGGGCGGAAGG
- Phenotype
-
MIM: 124015
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4651
PubMed Links
Gene Details
- Gene
- MIR4651
- Gene Name
- microRNA 4651
There are no transcripts associated with this gene.
- Gene
- POR
- Gene Name
- cytochrome p450 oxidoreductase
There are no transcripts associated with this gene.
View Full Product Details