Product Details

SNP ID
rs3818094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:44899482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAGCAATATTAGCTATGCTACT[A/G]TTCACTTAAGATTGATTTTCATGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf28 PubMed Links

Gene Details

Gene
C14orf28
Gene Name
chromosome 14 open reading frame 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017923.1 Intron NP_001017923.1
XM_011536408.1 Intron XP_011534710.1
Gene
LOC101927418
Gene Name
uncharacterized LOC101927418
There are no transcripts associated with this gene.

View Full Product Details