Product Details

SNP ID
rs3738673
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:39739151 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GCAGACTTCTTAGCAGTACGCACTT[C/T]TCCAGCTTCTTTGCTTTGCTTCCCG
Phenotype
MIM: 602435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIE PubMed Links

Gene Details

Gene
PPIE
Gene Name
peptidylprolyl isomerase E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195007.1 Intron NP_001181936.1
NM_001319293.1 Intron NP_001306222.1
NM_006112.3 Intron NP_006103.1
NM_203456.2 Intron NP_982281.1
XM_006710289.2 Intron XP_006710352.1
XM_006710290.3 Intron XP_006710353.1
XM_011540501.1 Intron XP_011538803.1
XM_017000051.1 Intron XP_016855540.1
XM_017000052.1 Intron XP_016855541.1

View Full Product Details