Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001045556.2 | 1344 | UTR 3 | NP_001039021.1 | ||
NM_001045557.2 | 1344 | UTR 3 | NP_001039022.2 | ||
NM_001282964.1 | 1344 | UTR 3 | NP_001269893.1 | ||
NM_001282965.1 | 1344 | UTR 3 | NP_001269894.1 | ||
NM_006748.3 | 1344 | UTR 3 | NP_006739.2 | ||
XM_017013739.1 | 1344 | UTR 3 | XP_016869228.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003235.4 | 1344 | Intron | NP_003226.4 | ||
XM_005251038.4 | 1344 | Intron | XP_005251095.1 | ||
XM_005251040.4 | 1344 | Intron | XP_005251097.1 | ||
XM_005251042.4 | 1344 | Intron | XP_005251099.1 | ||
XM_006716622.3 | 1344 | Intron | XP_006716685.1 | ||
XM_017013793.1 | 1344 | Intron | XP_016869282.1 | ||
XM_017013794.1 | 1344 | Intron | XP_016869283.1 | ||
XM_017013795.1 | 1344 | Intron | XP_016869284.1 | ||
XM_017013796.1 | 1344 | Intron | XP_016869285.1 | ||
XM_017013797.1 | 1344 | Intron | XP_016869286.1 | ||
XM_017013798.1 | 1344 | Intron | XP_016869287.1 | ||
XM_017013799.1 | 1344 | Intron | XP_016869288.1 | ||
XM_017013800.1 | 1344 | Intron | XP_016869289.1 |