Product Details

SNP ID
rs3740138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:109871173 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAGAGGCCGTATCAGACATGGA[C/T]GTTCCCTTGCCAGCTCTGCGACCCT
Phenotype
MIM: 602443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XPNPEP1 PubMed Links

Gene Details

Gene
XPNPEP1
Gene Name
X-prolyl aminopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167604.1 Intron NP_001161076.1
NM_001324128.1 Intron NP_001311057.1
NM_001324131.1 Intron NP_001311060.1
NM_001324132.1 Intron NP_001311061.1
NM_001324133.1 Intron NP_001311062.1
NM_001324134.1 Intron NP_001311063.1
NM_001324135.1 Intron NP_001311064.1
NM_001324136.1 Intron NP_001311065.1
NM_020383.3 Intron NP_065116.3
XM_011540134.1 Intron XP_011538436.1
XM_017016613.1 Intron XP_016872102.1
XM_017016614.1 Intron XP_016872103.1
XM_017016615.1 Intron XP_016872104.1

View Full Product Details