Product Details

SNP ID
rs3811036
Assay Type
Functionally Tested
NCBI dbSNP Submissions
52
Location
Chr.1:157515485 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
ATTAAAAAACCTGCCAGTCAGGGCT[G/T]TAACTGGGAAACAGGTGACAGTCCA
Phenotype
MIM: 605877
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
FCRL5 PubMed Links
Additional Information
For this assay, SNP(s) [rs75090424] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FCRL5
Gene Name
Fc receptor like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195388.1 3272 UTR 3 NP_001182317.1
NM_031281.2 3272 UTR 3 NP_112571.2
XM_011510030.1 3272 Intron XP_011508332.1
XM_011510031.1 3272 Intron XP_011508333.1
XM_011510032.1 3272 Intron XP_011508334.1
XM_011510033.1 3272 Intron XP_011508335.1

View Full Product Details