Product Details

SNP ID
rs3748133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76505264 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGTGCCAGGGAGTGGATGAGTCA[T/C]CTGGGAGGGGCTGGGATGGGAAGAA
Phenotype
MIM: 613141
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DTX2 PubMed Links

Gene Details

Gene
DTX2
Gene Name
deltex E3 ubiquitin ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102594.1 Intron NP_001096064.1
NM_001102595.1 Intron NP_001096065.1
NM_001102596.1 Intron NP_001096066.1
NM_020892.2 Intron NP_065943.2
XM_005250131.1 Intron XP_005250188.1
XM_005250132.1 Intron XP_005250189.1
XM_011515772.1 Intron XP_011514074.1
XM_011515773.2 Intron XP_011514075.1
XM_011515774.1 Intron XP_011514076.1
XM_011515775.1 Intron XP_011514077.1
XM_017011723.1 Intron XP_016867212.1
XM_017011724.1 Intron XP_016867213.1
XM_017011725.1 Intron XP_016867214.1
XM_017011726.1 Intron XP_016867215.1
XM_017011727.1 Intron XP_016867216.1
XM_017011728.1 Intron XP_016867217.1
XM_017011729.1 Intron XP_016867218.1
XM_017011730.1 Intron XP_016867219.1
Gene
LOC105375355
Gene Name
uroplakin-3b
There are no transcripts associated with this gene.

View Full Product Details