Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001267843.1 | 10421 | UTR 3 | NP_001254772.1 | ||
NM_004898.3 | 10421 | UTR 3 | NP_004889.1 | ||
XM_005265787.2 | 10421 | Intron | XP_005265844.1 | ||
XM_011534409.2 | 10421 | Intron | XP_011532711.1 | ||
XM_011534410.2 | 10421 | Intron | XP_011532712.1 | ||
XM_011534411.2 | 10421 | Intron | XP_011532713.1 | ||
XM_017008854.1 | 10421 | Intron | XP_016864343.1 | ||
XM_017008855.1 | 10421 | Intron | XP_016864344.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018475.4 | 10421 | Intron | NP_060945.2 | ||
XM_011534394.2 | 10421 | Intron | XP_011532696.1 | ||
XM_017008412.1 | 10421 | Intron | XP_016863901.1 |