Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_173348.1 | Intron | NP_775483.1 | |||
XM_005269744.2 | Intron | XP_005269801.1 | |||
XM_005269745.2 | Intron | XP_005269802.1 | |||
XM_005269747.3 | Intron | XP_005269804.1 | |||
XM_011539737.2 | Intron | XP_011538039.1 | |||
XM_011539740.2 | Intron | XP_011538042.1 | |||
XM_017016164.1 | Intron | XP_016871653.1 | |||
XM_017016165.1 | Intron | XP_016871654.1 | |||
XM_017016166.1 | Intron | XP_016871655.1 | |||
XM_017016167.1 | Intron | XP_016871656.1 |