Product Details

SNP ID
rs3822848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159970677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACCCTAAGATACATAGTTACTC[C/T]AGTAACTAGTATGATACAGAGAAAT
Phenotype
MIM: 147280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGF2R PubMed Links

Gene Details

Gene
IGF2R
Gene Name
insulin like growth factor 2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000876.3 Intron NP_000867.2

View Full Product Details