Product Details
- SNP ID
-
rs3823213
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:39336094 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTCTGGGCTTCCAAACCAGAGCA[C/T]GGGCCAAGGGAGAGAGCTGGCAAAT
- Phenotype
-
MIM: 613919
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KIF6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116706958] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIF6
- Gene Name
- kinesin family member 6
View Full Product Details