Product Details

SNP ID
rs3823864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44883989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCGCAACAGTGAGGCCTAGAGA[C/G]ATCTTTTATGCCAGATTACCCAGGT
Phenotype
MIM: 608887
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR4657 PubMed Links

Gene Details

Gene
MIR4657
Gene Name
microRNA 4657
There are no transcripts associated with this gene.

Gene
PURB
Gene Name
purine rich element binding protein B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033224.4 1397 UTR 3 NP_150093.1

View Full Product Details