Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001369.2 | 10919 | UTR 3 | NP_001360.1 | ||
XM_005248262.3 | 10919 | UTR 3 | XP_005248319.2 | ||
XM_017009177.1 | 10919 | UTR 3 | XP_016864666.1 | ||
XM_017009178.1 | 10919 | UTR 3 | XP_016864667.1 | ||
XM_017009179.1 | 10919 | UTR 3 | XP_016864668.1 | ||
XM_017009180.1 | 10919 | Intron | XP_016864669.1 | ||
XM_017009181.1 | 10919 | Intron | XP_016864670.1 | ||
XM_017009182.1 | 10919 | Intron | XP_016864671.1 | ||
XM_017009183.1 | 10919 | Intron | XP_016864672.1 | ||
XM_017009184.1 | 10919 | Intron | XP_016864673.1 | ||
XM_017009185.1 | 10919 | UTR 3 | XP_016864674.1 | ||
XM_017009186.1 | 10919 | UTR 3 | XP_016864675.1 | ||
XM_017009187.1 | 10919 | Intron | XP_016864676.1 | ||
XM_017009188.1 | 10919 | UTR 3 | XP_016864677.1 |