Product Details

SNP ID
rs3779995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66922980 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTTTGTCTCATTAGTACCTAGTA[A/T]TTCCACAAATGAGCTTATTCCAAAC
Phenotype
MIM: 612215 MIM: 612216
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MCMDC2 PubMed Links

Gene Details

Gene
MCMDC2
Gene Name
minichromosome maintenance domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136160.1 Intron NP_001129632.1
NM_001136161.1 Intron NP_001129633.1
NM_173518.4 Intron NP_775789.3
XM_005251174.2 Intron XP_005251231.1
XM_006716427.3 Intron XP_006716490.1
XM_006716429.2 Intron XP_006716492.1
XM_006716433.3 Intron XP_006716496.1
XM_011517467.2 Intron XP_011515769.1
XM_011517468.2 Intron XP_011515770.1
XM_011517469.2 Intron XP_011515771.1
XM_017013140.1 Intron XP_016868629.1
Gene
SNHG6
Gene Name
small nucleolar RNA host gene 6
There are no transcripts associated with this gene.

Gene
SNORD87
Gene Name
small nucleolar RNA, C/D box 87
There are no transcripts associated with this gene.

View Full Product Details