Product Details

SNP ID
rs3789041
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:160208686 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CCCAGGTACAACTGTTGATCAGTGA[G/T]AATTGAGAGCAGTTCCCCTAAACAA
Phenotype
MIM: 114250 MIM: 615820 MIM: 603434
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CASQ1 PubMed Links

Gene Details

Gene
CASQ1
Gene Name
calsequestrin 1
There are no transcripts associated with this gene.

Gene
DCAF8
Gene Name
DDB1 and CUL4 associated factor 8
There are no transcripts associated with this gene.

Gene
LOC729867
Gene Name
uncharacterized LOC729867
There are no transcripts associated with this gene.

Gene
PEA15
Gene Name
phosphoprotein enriched in astrocytes 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297576.1 Intron NP_001284505.1
NM_001297577.1 Intron NP_001284506.1
NM_001297578.1 Intron NP_001284507.1
NM_003768.4 Intron NP_003759.1
XM_006711599.2 Intron XP_006711662.1

View Full Product Details