Product Details

SNP ID
rs3807315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128741432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTAATAATTTGTATTAGAATGA[C/T]ATGTAGACATTCTTCCAGAAATGTC
Phenotype
MIM: 603420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALU PubMed Links

Gene Details

Gene
CALU
Gene Name
calumenin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130674.2 Intron NP_001124146.1
NM_001199671.1 Intron NP_001186600.1
NM_001199672.1 Intron NP_001186601.1
NM_001199673.1 Intron NP_001186602.1
NM_001219.4 Intron NP_001210.1
XM_017012659.1 Intron XP_016868148.1
Gene
FAM71F1
Gene Name
family with sequence similarity 71 member F1
There are no transcripts associated with this gene.

View Full Product Details