Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001159397.1 | Intron | NP_001152869.1 | |||
NM_001159398.1 | Intron | NP_001152870.1 | |||
NM_052938.4 | Intron | NP_443170.1 | |||
XM_005244866.2 | Intron | XP_005244923.1 | |||
XM_005244867.2 | Intron | XP_005244924.1 | |||
XM_005244869.3 | Intron | XP_005244926.1 | |||
XM_011509125.1 | Intron | XP_011507427.1 | |||
XM_011509126.1 | Intron | XP_011507428.1 | |||
XM_011509127.2 | Intron | XP_011507429.1 | |||
XM_011509128.1 | Intron | XP_011507430.1 | |||
XM_011509129.1 | Intron | XP_011507431.1 | |||
XM_011509130.1 | Intron | XP_011507432.1 | |||
XM_011509131.1 | Intron | XP_011507433.1 | |||
XM_011509132.2 | Intron | XP_011507434.1 | |||
XM_011509133.2 | Intron | XP_011507435.1 | |||
XM_011509134.2 | Intron | XP_011507436.1 | |||
XM_011509135.2 | Intron | XP_011507437.1 | |||
XM_011509136.1 | Intron | XP_011507438.1 | |||
XM_011509137.2 | Intron | XP_011507439.1 | |||
XM_017000227.1 | Intron | XP_016855716.1 |