Product Details

SNP ID
rs3823641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100090597 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATTGTCCCTCTCTTCCAGGTGTG[C/T]GAGATCATCGAGAGCCCCCTCTTTC
Phenotype
MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984 MIM: 194510
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPS6 PubMed Links

Gene Details

Gene
COPS6
Gene Name
COP9 signalosome subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006833.4 460 Silent Mutation TGC,TGT C143C NP_006824.2
Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
There are no transcripts associated with this gene.

Gene
MIR106B
Gene Name
microRNA 106b
There are no transcripts associated with this gene.

Gene
MIR25
Gene Name
microRNA 25
There are no transcripts associated with this gene.

Gene
MIR93
Gene Name
microRNA 93
There are no transcripts associated with this gene.

Gene
ZNF3
Gene Name
zinc finger protein 3
There are no transcripts associated with this gene.

View Full Product Details