Product Details
- SNP ID
-
rs3829759
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:132166779 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCCGCGAGTCCCGCCTGACCCC[A/G]TCGCTGCCTCTCCAGGGCTTCTCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NTNG2
PubMed Links
Gene Details
- Gene
- NTNG2
- Gene Name
- netrin G2
View Full Product Details