Product Details

SNP ID
rs3208122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2131361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAATCTTAGCGGGACTTTGGCCT[G/T]ATCCATACAGATATCGTAGTTGCTT
Phenotype
MIM: 147470 MIM: 610146
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IGF2 PubMed Links

Gene Details

Gene
IGF2
Gene Name
insulin like growth factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000612.5 2272 UTR 3 NP_000603.1
NM_001007139.5 2272 UTR 3 NP_001007140.2
NM_001127598.2 2272 UTR 3 NP_001121070.1
NM_001291861.2 2272 UTR 3 NP_001278790.1
NM_001291862.2 2272 UTR 3 NP_001278791.1
Gene
IGF2-AS
Gene Name
IGF2 antisense RNA
There are no transcripts associated with this gene.

Gene
INS-IGF2
Gene Name
INS-IGF2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042376.2 2272 Intron NP_001035835.1
Gene
MIR483
Gene Name
microRNA 483
There are no transcripts associated with this gene.

View Full Product Details