Product Details

SNP ID
rs3171487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44849702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGCTGCTCTTAGTGTATTTCA[A/G]AAAAAAGCAGATGGACGAAGACCGA
Phenotype
MIM: 603784
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTTG1IP PubMed Links

Gene Details

Gene
PTTG1IP
Gene Name
pituitary tumor-transforming 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286822.1 2709 UTR 3 NP_001273751.1
NM_004339.3 2709 UTR 3 NP_004330.1

View Full Product Details