Product Details

SNP ID
rs3730284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159917439 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTCTTGTCCACCGTCATCTCCAT[C/T]GGGCCTCTCCTTGGGTGGAAGGAGC
Phenotype
MIM: 104220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADRA1B PubMed Links

Gene Details

Gene
ADRA1B
Gene Name
adrenoceptor alpha 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000679.3 707 Silent Mutation ATC,ATT I178I NP_000670.1
XM_005265818.3 707 Silent Mutation ATC,ATT I178I XP_005265875.1
XM_005265819.2 707 Silent Mutation ATC,ATT I178I XP_005265876.1
XM_006714821.3 707 Silent Mutation ATC,ATT I178I XP_006714884.1
XM_011534435.1 707 Silent Mutation ATC,ATT I178I XP_011532737.1
XM_011534437.2 707 Silent Mutation ATC,ATT I178I XP_011532739.1
XM_011534438.2 707 Silent Mutation ATC,ATT I178I XP_011532740.1

View Full Product Details