Product Details

SNP ID
rs3742851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70642635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCGGTGCAGATTCTCCTGCTGT[C/G]ACTGTGTTGCTCTGGGGAGAACCCG
Phenotype
MIM: 610488
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928075 PubMed Links

Gene Details

Gene
LOC101928075
Gene Name
uncharacterized LOC101928075
There are no transcripts associated with this gene.

Gene
TTC9
Gene Name
tetratricopeptide repeat domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015351.1 Intron NP_056166.1

View Full Product Details