Product Details

SNP ID
rs3219177
Assay Type
Functionally Tested
NCBI dbSNP Submissions
40
Location
Chr.19:7669483 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
ACTTGGGCAAGCTCCCCAAGGGTCT[C/T]AGAGACCTCACTGATCCCTGGCACA
Phenotype
MIM: 609565 MIM: 605565
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MCEMP1 PubMed Links

Gene Details

Gene
MCEMP1
Gene Name
mast cell expressed membrane protein 1
There are no transcripts associated with this gene.

Gene
RETN
Gene Name
resistin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193374.1 Intron NP_001180303.1
NM_020415.3 Intron NP_065148.1

View Full Product Details