Product Details

SNP ID
rs3747444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149541156 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGCTTGGGAGCCCATCGTCCTG[G/T]CTCCGGTGGCCTCGCTGGGTCTCGG
Phenotype
MIM: 300954
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CXorf40A PubMed Links

Gene Details

Gene
CXorf40A
Gene Name
chromosome X open reading frame 40A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171907.2 183 Intron NP_001165378.1
NM_001171908.2 183 Intron NP_001165379.1
NM_001171909.3 183 Intron NP_001165380.1
NM_001324274.1 183 Intron NP_001311203.1
NM_001324275.1 183 Intron NP_001311204.1
NM_001324276.1 183 UTR 5 NP_001311205.1
NM_001324277.1 183 Intron NP_001311206.1
NM_001324278.1 183 Intron NP_001311207.1
NM_001324279.1 183 UTR 5 NP_001311208.1
NM_001324280.1 183 Intron NP_001311209.1
NM_178124.5 183 Intron NP_835225.2
XM_005262349.4 183 Intron XP_005262406.2
Gene
LINC00893
Gene Name
long intergenic non-protein coding RNA 893
There are no transcripts associated with this gene.

Gene
LOC101928917
Gene Name
heat shock transcription factor, X-linked-like
There are no transcripts associated with this gene.

View Full Product Details