Product Details

SNP ID
rs3749872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107067300 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTTCCAGGACTCTGGTATGACA[C/T]ACTGTTTGCATTCGACTGTTTCCTT
Phenotype
MIM: 616374
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BEND3 PubMed Links

Gene Details

Gene
BEND3
Gene Name
BEN domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080450.2 4541 UTR 3 NP_001073919.1
XM_005267079.3 4541 Intron XP_005267136.1
XM_005267080.3 4541 UTR 3 XP_005267137.1
XM_011536005.2 4541 Intron XP_011534307.1
XM_017011138.1 4541 UTR 3 XP_016866627.1

View Full Product Details