Product Details

SNP ID
rs3759983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69107235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTGATTGCACGTGGGGTATCTGG[C/T]TGAGGGACATTTTGGGGGCCTCTAT
Phenotype
MIM: 602428
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAS3 PubMed Links

Gene Details

Gene
HAS3
Gene Name
hyaluronan synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199280.1 Intron NP_001186209.1
NM_005329.2 Intron NP_005320.2
NM_138612.2 Intron NP_619515.1
XM_005255921.2 Intron XP_005255978.1
XM_011523061.2 Intron XP_011521363.1

View Full Product Details