Product Details

SNP ID
rs3764905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66633239 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGCAGCCCCAGCTCTCATTCCCC[G/A]TTGCCTGCCTGAGAGCCCTGGTGAT
Phenotype
MIM: 611725
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
KCTD7 PubMed Links

Gene Details

Gene
KCTD7
Gene Name
potassium channel tetramerization domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167961.2 Intron NP_001161433.1
NM_153033.4 Intron NP_694578.1

View Full Product Details