Product Details

SNP ID
rs3778485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46659208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTGAGCATTTACTGCAGATGTA[C/T]ACAGACATACATGTATATACATATA
Phenotype
MIM: 605994 MIM: 613725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP39A1 PubMed Links

Gene Details

Gene
CYP39A1
Gene Name
cytochrome P450 family 39 subfamily A member 1
There are no transcripts associated with this gene.

Gene
SLC25A27
Gene Name
solute carrier family 25 member 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204051.1 Intron NP_001190980.1
NM_001204052.1 Intron NP_001190981.1
NM_004277.4 Intron NP_004268.3
XM_005249483.2 Intron XP_005249540.1
XM_017011512.1 Intron XP_016867001.1

View Full Product Details