Product Details

SNP ID
rs3799843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109701348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACAGTCATACAGCCCAGTGCAA[C/G]TAGGAGTTTCCAGCAGAGATTCAGC
Phenotype
MIM: 609390
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FIG4 PubMed Links

Gene Details

Gene
FIG4
Gene Name
FIG4 phosphoinositide 5-phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014845.5 439 Intron NP_055660.1
XM_011536281.2 439 UTR 5 XP_011534583.1
XM_017011591.1 439 Intron XP_016867080.1
XM_017011592.1 439 Intron XP_016867081.1
XM_017011593.1 439 Intron XP_016867082.1

View Full Product Details