Product Details

SNP ID
rs3810649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152637947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCGCACCTCGCAGCTGCCGGGCG[A/G]GCCCTGGGGGGAGCTGCGTCCAGCA
Phenotype
MIM: 300349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRQ PubMed Links

Gene Details

Gene
GABRQ
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018558.3 53 Intron NP_061028.3
XM_011531184.2 53 UTR 5 XP_011529486.1

View Full Product Details