Product Details

SNP ID
rs3820382
Assay Type
Functionally Tested
NCBI dbSNP Submissions
6
Location
Chr.1:202133981 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CCTCTTCTGATCTGAACTTGCCCCA[A/G]GTCCACCTGCGTCTCCCTCCTTCTC
Phenotype
MIM: 616597
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARL8A PubMed Links

Gene Details

Gene
ARL8A
Gene Name
ADP ribosylation factor like GTPase 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256129.1 1150 UTR 3 NP_001243058.1
NM_138795.3 1150 UTR 3 NP_620150.1
Gene
GPR37L1
Gene Name
G protein-coupled receptor 37 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004767.3 1150 Intron NP_004758.3
XM_011510158.2 1150 Intron XP_011508460.1

View Full Product Details