Product Details
- SNP ID
-
rs104894958
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.Y:2787327 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCATTTTCCACTGGTATCCCAGCT[G/A]CTTGCTGATCTCTGAGTTTCGCATT
- Phenotype
-
MIM: 480000
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SRY
PubMed Links
Gene Details
- Gene
- SRY
- Gene Name
- sex determining region Y
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003140.2 |
415 |
Nonsense Mutation |
CAG,TAG |
Q93* |
NP_003131.1 |
View Full Product Details