Product Details

SNP ID
rs5030803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10142068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAACTCGCGCGAGCCCTCCCAGG[G/T]CATCTTCTGCAATCGCAGTCCGCGC
Phenotype
MIM: 608537
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VHL PubMed Links

Gene Details

Gene
VHL
Gene Name
von Hippel-Lindau tumor suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000551.3 434 Missense Mutation GGC,GTC G74V NP_000542.1
NM_198156.2 434 Missense Mutation GGC,GTC G74V NP_937799.1

View Full Product Details