Product Details
- SNP ID
-
hCV27536080
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:48684340 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGCTGCCCCCTGGAGCTGAGCAC[C/T]GGACCAAGGAGCATTGTGGGGCTGT
- Phenotype
-
MIM: 300392
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WAS
PubMed Links
Gene Details
- Gene
- WAS
- Gene Name
- Wiskott-Aldrich syndrome
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000377.2 |
247 |
Missense Mutation |
CGG,TGG |
R64W |
NP_000368.1 |
View Full Product Details