Product Details

SNP ID
rs104894873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:43949817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGCCTCAGGAATTGCATTCCTC[T/G]CAGTGACAGGAGAGGATGTACCGGT
Phenotype
MIM: 300658
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
NDP PubMed Links

Gene Details

Gene
NDP
Gene Name
NDP, norrin cystine knot growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000266.3 963 Silent Mutation NP_000257.1

View Full Product Details