Product Details

SNP ID
rs137853247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120978860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGGCACTGATCCAGGCACTGG[A/G]TGAGCCGGGGCCCTACCTCCTGGCT
Phenotype
MIM: 142410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNF1A PubMed Links

Gene Details

Gene
HNF1A
Gene Name
HNF1 homeobox A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000545.6 293 Missense Mutation GAT,GGT D31G NP_000536.5
NM_001306179.1 293 Missense Mutation GAT,GGT D31G NP_001293108.1
XM_005253931.3 293 Missense Mutation GAT,GGT D31G XP_005253988.1
Gene
HNF1A-AS1
Gene Name
HNF1A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details