Product Details
- SNP ID
-
hCV27540491
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:37783538 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGCAGCCTGCCTGAATTTCAAC[C/T]GCATGCTGATTCTCTTGCCAGTCTG
- Phenotype
-
MIM: 300481
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYBB
PubMed Links
Gene Details
- Gene
- CYBB
- Gene Name
- cytochrome b-245 beta chain
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000397.3 |
251 |
Missense Mutation |
CGC,TGC |
R64C |
NP_000388.2 |
View Full Product Details