Product Details

SNP ID
rs115117837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70884002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAAGCTGGCCAGGTTTATGTCC[C/T]GTTCTGAAAGGCCGGCACACTCATG
Phenotype
MIM: 126090 MIM: 603729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCBD1 PubMed Links

Gene Details

Gene
PCBD1
Gene Name
pterin-4 alpha-carbinolamine dehydratase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000281.3 662 Missense Mutation CAG,CGG Q88R NP_000272.1
NM_001289797.1 662 Missense Mutation CAG,CGG Q39R NP_001276726.1
NM_001323004.1 662 Intron NP_001309933.1
Gene
SGPL1
Gene Name
sphingosine-1-phosphate lyase 1
There are no transcripts associated with this gene.

View Full Product Details