Product Details

SNP ID
rs61753179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:89178060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAACCTGATGGAGAAGGAATGCT[A/G]TCCACCGTGGAGCGGGGACAGGAGT
Phenotype
MIM: 606933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TYR PubMed Links

Gene Details

Gene
TYR
Gene Name
tyrosinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000372.4 496 Missense Mutation TAT,TGT Y36C NP_000363.1
XM_011542970.2 496 Missense Mutation TAT,TGT Y36C XP_011541272.1

View Full Product Details